missing translation for 'onlineSavingsMsg'
Learn More

Sodium iodate, 99% min

Product Code. 11399277
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399277 500 g 500g
11389277 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399277 Supplier Thermo Scientific Alfa Aesar Supplier No. 040135.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7681-55-2 | INaO3 | 197.891 g/mol

Sodium iodate is used in animal feed, food industry. It is utilized in the manufacture of disinfectants, antiseptics and deodorants. It acts as a significant part in the oxidation of aromatic, aliphatic, and alicyclic thiols. Potassium tetrahydroborate and sodium iodate in acidic aqueous solution react with cadmium to generate chemical vapor, which is useful for atomic fluorescence spectrometry application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-55-2
Molecular Formula INaO3
Molecular Weight (g/mol) 197.891
MDL Number MFCD00003533
InChI Key WTCBONOLBHEDIL-UHFFFAOYSA-M
Synonym sodium iodate, natriumjodat german, iodic acid, sodium salt, sodium iodate naio3, iodic acid hio3 , sodium salt, sodium iodate naio3 pentahydrate, unii-u558pcs5z9, hsdb 749, iodic acid, sodium salt, pentahydrate, natriumjodat
PubChem CID 23675764
ChEBI CHEBI:81708
IUPAC Name sodium;iodate
SMILES [O-]I(=O)=O.[Na+]

Specifications

Melting Point (decomposition)
Color White
Physical Form Crystalline Powder
Quantity 500 g
UN Number UN1479
Merck Index 14,8630
Solubility Information Soluble in water and acetone.
Formula Weight 197.89
Percent Purity ≤99%
Sensitivity Light and moisture sensitive
Density 4.28 g/mL
Chemical Name or Material Sodium iodate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium iodate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Oxidising solid Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
Supplemental information
  • MIXTURE LIST-Contain: Sodium iodate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.