missing translation for 'onlineSavingsMsg'
Learn More

Sodium iodate, 99%, extra pure, reagent, powder

Product Code. 10697381 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle
Unit Size:
500g
5g
This item is not returnable. View return policy

Product Code. 10697381

Brand: Thermo Scientific Acros 419565000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7681-55-2 | INaO3 | 197.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-55-2
Molecular Formula INaO3
Molecular Weight (g/mol) 197.89
MDL Number MFCD00003533
InChI Key WTCBONOLBHEDIL-UHFFFAOYSA-M
Synonym sodium iodate, natriumjodat german, iodic acid, sodium salt, sodium iodate naio3, iodic acid hio3 , sodium salt, sodium iodate naio3 pentahydrate, unii-u558pcs5z9, hsdb 749, iodic acid, sodium salt, pentahydrate, natriumjodat
PubChem CID 23675764
ChEBI CHEBI:81708
IUPAC Name sodium;iodate
SMILES [O-]I(=O)=O.[Na+]

Specifications

Melting Point 560.0°C
Color White
pH 5.0 to 8.0 (5% soln. at 20°C)
Physical Form Fine Crystalline Powder
Quantity 500 g
Assay Percent Range 98.5% min. (Iodometry)
Linear Formula NaIO3
Merck Index 15, 8764
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone, insoluble in alcohol
Formula Weight 197.89
Percent Purity 99%
Grade Extra Pure
Packaging Glass bottle
Chemical Name or Material Sodium iodate, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium iodate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.