missing translation for 'onlineSavingsMsg'
Learn More

Sodium ingot, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog Number 11341428
Click to view available options
Quantity:
450 g
5 x 450 g
7440-23-5
Na
22.99
MFCD00085307
KEAYESYHFKHZAL-UHFFFAOYSA-N
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
5360545
CHEBI:26708
sodium
[Na]
7440-23-5
Na
22.99
MFCD00085307
KEAYESYHFKHZAL-UHFFFAOYSA-N
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
5360545
CHEBI:26708
sodium
[Na]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver to Gray
Appearance Silver Gray ingot
Density 0.968g/cm3
Assay Percent Range 99.8% (Metals basis)
Quantity 450 g
Grade ACS
Odor Odorless
Vapor Pressure 1.59 hPa (1mm Hg)
Form Ingot
Chemical Name or Material Sodium, Product meets ACS specifications

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'