missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite Strong Solution According to European Pharmacopoeia (EP), Reagecon

Product Code. 12802503 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12802503 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12802503 Supplier Reagecon Supplier No. 10816005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity stable product developed and tested for European Pharmacopoeia (EP) applications including use as a reagent in the performance of tests outlined in several EP monographs.

  • European Pharmacopoeia Reagent, Solution
  • High-purity stable product developed and tested for European Pharmacopoeia (EP) applications.
  • These include use as a reagent in the performance of tests outlined in several EP monographs.
  • This Reagecon EP reagent is accurate, stable and reproducible.
  • Stability is proven (either opened or unopened) for the shelf life of this product.

Specifications

Packaging Amber Glass Winchester Bottle GL45
Physical Form Liquid
Technique EP Testing
Quantity 500 mL
compliance-icons compliance-icons
Product Identifier
  • SODIUM HYPOCHLORITE SOLUTION, STRONG
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H400-Very toxic to aquatic life.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: sodium hypochlorite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.