missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite Solution, TRC

Product Code. 30415348
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30415348 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30415348 Supplier Toronto Research Chemicals Supplier No. TRCS6349855ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium hypochlorite
CAS 7681-52-9
Molecular Formula Cl O . Na
Quantity 5 mL
Synonym Hypochlorous Acid Sodium Salt, AD Gel, ActiPlus N 2818, AgClor 310, Antiformin, Aron Clean, Aron Clean LB 10, B-K Liquid, Baso Agri+, Belizna, Belkina, Carrel-Dakin Solution, Chloros, Cloralex, Cloralex NL, Clorox, Comfor, Dakin's Solution, DanKlorix, Deosan, Dispatch, Dispatch (Salt), Fox-Chlor, Hishikurin S, Histolith, Hyclorite, Hypex, Hypure, Hypure N, Javel Water, Javelle Water, Javex, Javex 5, Jomax Mold & Mildew, Stain Remover, Klorocin, Linely, Milton, Milton Sterilising Fluid, Modified Dakin's Solution, Nades, Neo-cleaner, Neoseptal CL, Parazone, Parozone, Purelex, Purelox, Purin B, Raverrack D, Sanrack P, Sodium Chloride Oxide (NaClO), Sodium Hypochloride, Sodium Hypochlorite (NaClO), Sodium Hypochlorite (NaOCl)
InChI Formula InChI=1S/ClO.Na/c1-2;/q-1;+1
SMILES [Na+].[O-]Cl
IUPAC Name sodium;hypochlorite
Molecular Weight (g/mol) 74.442
Formula Weight 73.954
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.