missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite Solution, Reagent grade, Available chlorine 4.00 to 4.99%, Honeywell

Product Code. p-7071927 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
25 mL
500 mL
3 L
5 L
Unit Size:
25mL
3 L
5 L
500mL
This item is not returnable. View return policy

Product Code. 15638340

Brand: Honeywell Research Chemicals 23930525ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, available chlorine 4.00-4.99 %

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-52-9
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.439
MDL Number MFCD00011120
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146
IUPAC Name sodium;hypochlorite
SMILES [O-]Cl.[Na+]

Specifications

Quantity 25 mL
Linear Formula NaClO
UN Number 3082
Formula Weight 74.44g/mol
Grade Reagent
Concentration or Composition (by Analyte or Components) Available chlorine 4.00 to 4.99%
Vapor Pressure 17.5mmHg (20°C)
Density 1.097g/mL at 25°C
Chemical Name or Material Sodium Hypochlorite Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.