missing translation for 'onlineSavingsMsg'
Learn More

Sodium hypochlorite solution, Purum, ∼10% (RT), Honeywell Fluka™

Product Code. 16685913 Shop All Honeywell Fluka Products
Click to view available options
:
25 L
This item is not returnable. View return policy

Product Code. 16685913

Brand: Honeywell Fluka 716962.5L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
  • ≥95% for general laboratory use
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 111°C
Chemical Name or Material Sodium hypochlorite solution
Melting Point -30°C to -20°C
CAS 7732-18-5, 7681-52-9
Color Clear
Physical Form Liquid
Quantity 25 L
Molecular Formula NaClO
UN Number UN1791
Molecular Weight (g/mol) 74.44 g/mol
Grade Synthesis
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.