missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hypochlorite Solution, 5% Available Chlorine, BAKER ANALYZED™ Reagent, J.T. Baker™
Shop All J.T.Baker ProductsDescription
- For Laboratory,Research,or Manufacturing Use
Specifications
Specifications
| Quantity | 4 L |
| Formula Weight | 74.44 |
| Percent Purity | 5% (Available Chlorine) |
| Grade | BAKER ANALYZED™ Reagent |
| Packaging | Pressure Bottom Poly Bottle |
| Specific Gravity | 1L = 1.10kg |
| Chemical Name or Material | Sodium Hypochlorite Solution |
Safety and Handling
Product Identifier
- Sodium Hypochlorite 5% Solution
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Sodium hypochlorite
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?