missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite Solution, 47/50%, Chem-Lab

Product Code. 11909012
Change view
Click to view available options
Quantity:
1 L
5 L
Unit Size:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11909012 1 L 1 L
11919012 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11909012 Supplier Chem-Lab Supplier No. CL00.1410.1000
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used as a disinfectant and bleaching agent. Sodium Hypochlorite Solution, 47/50%, Chem-Lab is a chemical compound also known as bleach. It is a common laboratory reagent that reacts with many acids and is a strong oxidizer.

Store in a cool environment, at 2–15°C.

Chemical Identifiers

CAS 7681-52-9
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.439
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146
IUPAC Name sodium;hypochlorite
SMILES [O-]Cl.[Na+]

Specifications

Color Yellow
Boiling Point 100°C
Melting Point 0°C
Composition Available chlorine: ± 13%
Density 1.22g/mL
Solubility Information Water Soluble
Formula Weight 74.44g/mol
Percent Purity 47/50% (NaClO)
Odor Chlorine
pH >13
Quantity 1 L
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hypochlorite 47/50% solution p.
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.