Learn More
Sodium hypochlorite, 11-15% available chlorine
Description
Sodium hypochlorite is used as a disinfectant and a bleaching agent. It is used as a destainer involved in removing of mold stains, dental stains arises by fluorosis. It finds application in laundry detergents. Its aqueous solution is used as household bleach for sanitizing smooth surfaces. In a water treatment plant, it is utilized for treating dilute cyanide waste water like electroplating wastes. It is an oxidizing agent used with phase transfer catalyst to convert alcohols into corresponding carbonyl compounds. It is an important reactant in Hofmann rearrangement to prepare primary amine from primary amide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Yellow |
| Physical Form | Liquid |
| Quantity | 2.5 L |
| Solubility Information | Miscible with water. |
| SMILES | [Na+].[O-]Cl |
| IUPAC Name | sodium hypochlorite |
| Molecular Weight (g/mol) | 74.44 |
| Formula Weight | 74.44 |
| Density | 1.2 |
| Chemical Name or Material | Sodium Hypochlorite aq. soln., 11-15% available chlorine |
Safety and Handling
- Sodium hypochlorite, 11-15% available chlorine
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
- EUH031-Contact with acids liberates toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Sodium hypochlorite,Sodium hydroxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.