missing translation for 'onlineSavingsMsg'
Learn More

Sodium hypochlorite, 11-15% available chlorine

Product Code. 15450837
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450837 2.5 L 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15450837 Supplier Thermo Scientific Alfa Aesar Supplier No. L14709.0F
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7681-52-9 ∣ ClNaO ∣ 74.439 g/mol

Sodium hypochlorite is used as a disinfectant and a bleaching agent. It is used as a destainer involved in removing of mold stains, dental stains arises by fluorosis. It finds application in laundry detergents. Its aqueous solution is used as household bleach for sanitizing smooth surfaces. In a water treatment plant, it is utilized for treating dilute cyanide waste water like electroplating wastes. It is an oxidizing agent used with phase transfer catalyst to convert alcohols into corresponding carbonyl compounds. It is an important reactant in Hofmann rearrangement to prepare primary amine from primary amide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-52-9
Molecular Formula ClNaO
MDL Number MFCD00011120
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146

Specifications

Color Yellow
Physical Form Liquid
Quantity 2.5 L
Density 1.2
Solubility Information Miscible with water.
SMILES [Na+].[O-]Cl
IUPAC Name sodium hypochlorite
Molecular Weight (g/mol) 74.44
Formula Weight 74.44
Grade Ungraded
Chemical Name or Material Sodium Hypochlorite aq. soln., 11-15% available chlorine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium hypochlorite, 11-15% available chlorine
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Sodium hypochlorite,Sodium hydroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.