missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide solution, Volumetric, 5.0 M NaOH (5.0N), Solstice

Product Code. 16626183
Change view
Click to view available options
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16626183 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16626183 Supplier Fluka Supplier No. 6560461L
  • ONLINE EXCLUSIVE PRICE
    £41.00
    £37.18 / 1 L
    Save £3.82
    9% Off
 Restricted Item
Estimated Shipment: 19-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemical Name or Material Sodium hydroxide solution
pH 14
Color Colorless
Boiling Point 105°C to 140°C (1,013 hPa)
CAS 1310-73-2
CAS Min % ≥ 70%
Melting Point -12°C to -10°C
CAS Max % <90%
Linear Formula NaOH
Density 1.327 g/cm3 (25°C)
Quantity 1 L
Molecular Weight (g/mol) 40.0 g/mol
Grade Volumetric Titration
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hydroxide solution
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.