missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Solution, Volumetric, 10.0M NaOH (10.0N), Honeywell™ Fluka™
Click to view available options
Quantity:
1 L
6 x 1 L
Packaging:
Poly Bottle
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
pH | >14 |
Color | Colorless |
Boiling Point | 120°C |
Linear Formula | NaOH |
UN Number | UN1824 |
Density | 1.327g/mL at 25°C |
Solubility Information | Soluble in water |
Quantity | 1 L |
Formula Weight | 40g/mol |
Packaging | Poly Bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'