missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide solution, Volumetric, 0.5 M NaOH (0.5N), Honeywell Fluka
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle
Unit Size:
1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
| Boiling Point | >100°C |
| CAS | 1310-73-2, 7732-18-5 |
| CAS Min % | ≥90.0000%, ≥1.0000% |
| CAS Max % | ≤100.0000%, <2.0000% |
| Molecular Formula | HNaO |
| Linear Formula | NaOH |
| Concentration | 0.5 M NaOH (0.5N) |
| MDL Number | MFCD00003548 |
| UN Number | UN1824 |
| Density | 1.020 g/cm3 |
| Show More |
Safety and Handling
Product Identifier
- Sodium hydroxide solution
- Warning
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : sodium hydroxide
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction