missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide solution, Volumetric, 0.5 M NaOH (0.5N), Solstice

Product Code. 16636183
Change view
Click to view available options
55 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16636183 55 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16636183 Supplier Fluka Supplier No. 7142455L
  • ONLINE EXCLUSIVE PRICE
    £919.00
    £846.92 / 55 L
    Save £72.08
    7% Off
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemical Name or Material Sodium hydroxide solution
Color Colorless
Boiling Point >100°C (1,013 hPa)
CAS 7732-18-5
CAS Min % ≥ 90%
CAS Max % ≤ 100%
Linear Formula NaOH
Density 1.020 g/cm3 (20°C)
Synonym NaOH
Quantity 55 L
Molecular Weight (g/mol) 40.00 g/mol
Grade Volumetric Titration
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.