missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution, TraceSELECT™, Honeywell™

Product Code. 15651270 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15651270 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15651270 Supplier Honeywell-Fluka Supplier No. 13171250ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1310-73-2
Molecular Formula HNaO
Linear Formula NaOH
MDL Number MFCD00003548
UN Number 1824
Density 1.335g/mL at 20°C
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 250 mL
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40g/mol
Percent Purity ≥30% (aq. soln.)
Grade TraceSELECT
Vapor Pressure 3mmHg (37°C)
Chemical Name or Material Sodium Hydroxide Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.