missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution, Strong, Reagecon™
BIOPHARMA

Product Code. 11474739 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474739 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11474739 Supplier Reagecon Supplier No. 1081404
  • ONLINE EXCLUSIVE PRICE
    £152.00
    £148.22 / 1 L
    Save £3.78
    2% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured and tested in compliance with the Chapter 4.1.1 of the European Pharmacopoeia.

Supplied with a Certificates of Analysis. Lot number and expiry date are stated on all product labels.

  • Molecular Formula: NaOH
  • Molar Mass: 40.00g/mol
  • CAS: 1310-73-2
  • EIN ECS No.: 215-185-5

Specifications

Color Colorless
Boiling Point 100°C
CAS 1310-73-2
Molecular Formula HNaO
Concentration 25–50%
Density 1.4746g/cm3
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Solubility Information Fully miscible
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 1 L
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40.00g/mol
Traceability to NIST Traceable to NIST
Vapor Pressure 23hPa (at 20°C)
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • SODIUM HYDROXIDE SOLUTION
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.