missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide solution, Volumetric, Reag. Ph. Eur., 1 M NaOH (1N), Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
5 L
10 L
20 L
4 x 5 L
6 x 1 L
Unit Size:
1 L
10 L
20 L
5 L
Pack of 4
Pack of 6
Packaging:
Plastic Bottle
VOLPAC™ CONTAINER
VOLPAK Container
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
| Linear Formula | NaOH |
| Concentration | 1M |
| UN Number | UN1824 |
| Quantity | 20 L |
| Formula Weight | 40g/mol |
| Vapor Pressure | 3 mmHg (37°C) |
| Chemical Name or Material | Sodium hydroxide solution |
Safety and Handling
Product Identifier
- Sodium hydroxide solution
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : sodium hydroxide; caustic soda
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction