missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide solution, Purum, ≥32%, Honeywell Fluka™

Product Code. 15690790 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
2.5 L
5 L
35 kg
4 x 2.5 L
4 x 5 L
Unit Size:
2.5 L
35kg
5 L
Pack of 4
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15690790 4 x 2.5 L Pack of 4
15670790 2.5 L 2.5 L
15610800 5 L 5 L
15600800 4 x 5 L Pack of 4
15680790 35 kg 35kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15690790 Supplier Honeywell-Fluka Supplier No. 052114X2.5L
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Purum,≥32%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Linear Formula NaOH
UN Number UN1824
Quantity 4 x 2.5 L
Formula Weight 40
Percent Purity ≥32%
Grade purum
Vapor Pressure 3mmHg (37°C)
Chemical Name or Material Sodium Hydroxide Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.