missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide solution, 50% in H2O, Honeywell

Product Code. 15668400 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
1 L
4 L
18 L
Unit Size:
1 L
100mL
18 L
25mL
4 L
500mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15668400 25 mL 25mL
15665460 100 mL 100mL
15695460 500 mL 500mL
15685460 1 L 1 L
15678400 4 L 4 L
15675460 18 L 18 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15668400 Supplier Honeywell Research Chemicals Supplier No. 41541325ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

50% in H2O

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Linear Formula NaOH
Concentration 50% in H2O
UN Number UN1824
Density 1.515g/mL at 25°C
Quantity 25 mL
Formula Weight 40g/mol
Vapor Pressure 3 mmHg (37°C)
Chemical Name or Material Sodium hydroxide solution
compliance-icons
Produktidentifikator
  • Sodium hydroxide solution
Signalord
  • Danger
Farekategori
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Faresætning
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Sikkerhedssætning
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplerende oplysninger
  • MIXTURE LIST-Contain: sodium hydroxide; caustic soda
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.