missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide solution, ≥32%, for determination of nitrogen, ≤0.0001% N, Honeywell™

Product Code. 15623060 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
4 x 5 L
6 x 1 L
Packaging:
Poly Bottle
Unit Size:
1 L
2.5 L
5 L
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 15623060

Brand: Honeywell Research Chemicals 305314X2.5L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

≥32%, for determination of nitrogen, ≤0.0001% N

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Boiling Point 120°C
CAS Min % ≥50.0000%, ≥25.0000%
CAS Max % <75.0000%, <50.0000%
Linear Formula NaOH
UN Number UN1824
Density 1.350 g/cm3
Quantity 4 x 2.5 L
Formula Weight 40.0
Percent Purity >32%
Packaging Poly Bottle
Chemical Name or Material Sodium hydroxide solution, ≥32%, for determination of nitrogen, ≤0.0001% N
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hydroxide solution
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.