missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution 10M (10N) ≈40 (w/v), Standard Solution Ready To Use, for Volumetric Analysis

Product Code. 10715721 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
25 L
Packaging:
Bag in box
HDPE plastic bottle
Plastic container
Unit Size:
1 L
10 L
2.5 L
25 L
5 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10715721 10 L Bag in box 10 L
10488790 1 L HDPE plastic bottle 1 L
10040220 2.5 L HDPE plastic bottle 2.5 L
10306240 5 L HDPE plastic bottle 5 L
10498980 25 L Plastic container 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10715721 Supplier Fisher Chemical Supplier No. J/7800/24
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium Hydroxide Solution 10M (10N) ≈40 (w/v), Standard Solution Ready To Use, for Volumetric Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number 3548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

pH 12
Boiling Point 120°C
Melting Point 8°C
Quantity 10 L
Packaging Bag in box
Physical Form Liquid
compliance-icons
Product Identifier
  • Sodium hydroxide solution 10M (10N) approx. 40% w/v
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P234-Keep only in original container.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.