missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, Solution 0.01N, for Volumetric Analysis, Fisher Chemical

Product Code. 12666727 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.50 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12666727 2.5 L 2.50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12666727 Supplier Fisher Chemical Supplier No. J/9010/17

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

pH 12
Color Colorless
Boiling Point 100°C
CAS 1310-73-2
Melting Point 0°C
Molecular Formula HNaO
MDL Number 3548
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 2.5 L
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Packaging Amber glass bottle
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.