missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, Reagent Grade, 97%, Powder, Honeywell™
Reagent Grade, powder, 97%
£34.01 - £163.70
Chemical Identifiers
CAS | 1310-73-2 |
---|---|
Molecular Formula | HNaO |
Molecular Weight (g/mol) | 39.997 |
MDL Number | MFCD00003548 |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
PubChem CID | 14798 |
ChEBI | CHEBI:32145 |
IUPAC Name | sodium;hydroxide |
SMILES | [OH-].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15686130
|
Honeywell Research Chemicals
655104-25G |
25 g | Poly Bottle | ||||||
15696130
|
Honeywell Research Chemicals
655104-500G |
500 g | Poly Bottle | ||||||
15676130
|
Honeywell Research Chemicals
655104-2.5KG |
2.5 kg | Poly Bottle | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
1310-73-2 | |
39.997 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
HNaO | |
MFCD00003548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
Specifications
Alkaline | |
1390°C | |
319°C | |
NaOH | |
UN1823 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
97% | |
>1 (vs air) | |
Solid |
Colorless | |
1310-73-2 | |
HNaO | |
MFCD00003548 | |
2.13g/cm3 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40g/mol | |
Reagent | |
18mmHg (20°C), 3mmHg (37°C) | |
Sodium hydroxide |
Safety and Handling
P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
missing translation for 'dotInformation' : 8 / PGII
missing translation for 'rtecsNumber' : 215-185-5