missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, Reagent Grade, 97%, Flakes, Honeywell™

Product Code. 15605660 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
10 kg
Packaging:
Drum with inliner
Poly Bottle
Unit Size:
10kg
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15605660 1 kg Poly Bottle 1kg
15695650 10 kg Drum with inliner 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15605660 Supplier Honeywell Research Chemicals Supplier No. 4840241KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, flakes, 97%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

pH Alkaline
Color Colorless
Boiling Point 1390°C
Melting Point 319°C
Linear Formula NaOH
UN Number UN1823
Density 2.13g/cm3 at 20°C
Quantity 1 kg
Formula Weight 40g/mol
Percent Purity 97%
Grade Reagent
Odor Odorless
Packaging Poly Bottle
Vapor Density >1 (vs air)
Vapor Pressure 3mmHg (37°C), 18mmHg (20°C)
Physical Form Solid
Chemical Name or Material Sodium hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.