missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, Puriss. p.a., ACS Reagent, K ≤0.02%, ≥98.0% (T), pellets, Solstice

Product Code. 16675913
Change view
Click to view available options
Quantity:
10 kg
Unit Size:
10kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16675913 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16675913 Supplier Fluka Supplier No. 7169010KG
  • ONLINE EXCLUSIVE PRICE
    £365.00
    £336.15 / 10kg
    Save £28.85
    7% Off
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Colorless
Boiling Point 1,390°C (1,013 hPa)
CAS 1310-73-2
Melting Point 319°C
CAS Max % 1
Molecular Formula HNaO
Linear Formula NaOH
Density 2.13 g/cm3 (20°C)
Synonym Caustic soda
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 10 kg
SMILES [OH-].[Na+]
IUPAC Name sodium hydroxide
Molecular Weight (g/mol) 40.00
Grade ACS
Chemical Name or Material Sodium hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.