missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, pearl, 97%

Product Code. 11465867
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
This item is not returnable. View return policy

Product Code. 11465867

Brand: Thermo Scientific Alfa Aesar B24414.0E

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1310-73-2 | HNaO | 39.997 g/mol

Sodium hydroxide, pearl is an important inorganic chemical used in various industries worldwide. In the paper industry, it is utilized in pulping and bleaching processes. It plays an important role in aluminum etching, chemical analysis and in paint strippers. It is an active component in a wide range of domestic products such as pipe and drain cleaners, oven cleaners and in home-cleaning products. Other uses include cleaning of fats, cleaning of equipment, ion exchange resin regeneration, pH control, degreasing agent for cleaning metal surface, de-enameling and scrubbing of off-gases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hydroxide, pearl is an important inorganic chemical used in various industries worldwide. In the paper industry, it is utilized in pulping and bleaching processes. It plays an important role in aluminum etching, chemical analysis and in paint strippers. It is an active component in a wide range of domestic products such as pipe and drain cleaners, oven cleaners and in home-cleaning products. Other uses include cleaning of fats, cleaning of equipment, ion exchange resin regeneration, pH control, degreasing agent for cleaning metal surface, de-enameling and scrubbing of off-gases.

Solubility
Soluble in water, methanol, glycerol and ethanol. Insoluble in acetone and ether.

Notes
Air sensitive and hygroscopic. It is deliquescent and readily absorbs moisture and carbon dioxide in air. Incompatible with strong oxidizing agents, strong acids and organic materials.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Boiling Point 1390°C
Melting Point 318°C
pH 14
Quantity 2500 g
UN Number UN1823
Merck Index 14,8627
Solubility Information Soluble in water,methanol,glycerol and ethanol. Insoluble in acetone and ether.
Formula Weight 40
Percent Purity 97%
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Density 2.13
Chemical Name or Material Sodium hydroxide, pearl
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hydroxide, pearl
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide, Disodium carbonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.