missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, MP Biomedicals

Product Code. 11488781 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g

Product Code. 11488781

Brand: MP Biomedicals 02153495.5

Please to purchase this item. Need a web account? Register with us today!

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Quantity 500 g
Formula Weight 40
Chemical Name or Material Sodium Hydroxide, Anhydrous
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.