missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide monohydrate, 99.996% (metals basis)

Product Code. p-4408654
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11369667

Brand: Thermo Scientific Alfa Aesar 041281.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12179-02-1 | H3NaO2 | 58.01 g/mol

Sodium hydroxide monohydrate is used as an acid neutralizer. It is a strong base widely used in the chemical industry to drive chemical reactions. It finds application as a neutralizing agent in the petroleum refining industry, as paint stripper and an analytical reagent in laboratories to measure the concentration of acids by titration. It is involved in soap production, alumina production by Bayer process, paper industry. It plays an important role as a catalyst for the transesterification of methanol and triglycerides in order to prepare biodiesel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12179-02-1
Molecular Formula H3NaO2
Molecular Weight (g/mol) 58.01
MDL Number MFCD00149229
InChI Key JBJWASZNUJCEKT-UHFFFAOYSA-M
Synonym sodium hydroxide monohydrate, na.ho.h2o, naoh water, water naoh, sodium hydroxide h2o, sodium hydroxide water, water sodium hydroxide, h2o naoh, naoh h2o, sodium,hydroxide,hydrate
PubChem CID 23719242
SMILES O.[OH-].[Na+]

Specifications

Color White
Boiling Point 1390°C
Melting Point 318.4°C
Assay Percent Range 99.996% (metals basis)
Density 2.13 g/cm3 at 20°C
Solubility Information Soluble in water and methanol.
Quantity 5 g
IUPAC Name sodium;hydroxide;hydrate
Formula Weight 58.01 (40.00 Anhydrous)
Odor Odorless
Physical Form Crystals
Chemical Name or Material Sodium hydroxide monohydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.