missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide monohydrate, 99.996% (metals basis)

Product Code. 11379667
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379667 100 g 100g
11369667 5 g 5g
11359667 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11379667 Supplier Thermo Scientific Alfa Aesar Supplier No. 041281.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12179-02-1 | H3NaO2 | 58.01 g/mol

Sodium hydroxide monohydrate is used as an acid neutralizer. It is a strong base widely used in the chemical industry to drive chemical reactions. It finds application as a neutralizing agent in the petroleum refining industry, as paint stripper and an analytical reagent in laboratories to measure the concentration of acids by titration. It is involved in soap production, alumina production by Bayer process, paper industry. It plays an important role as a catalyst for the transesterification of methanol and triglycerides in order to prepare biodiesel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12179-02-1
Molecular Formula H3NaO2
Molecular Weight (g/mol) 58.01
MDL Number MFCD00149229
InChI Key JBJWASZNUJCEKT-UHFFFAOYSA-M
Synonym sodium hydroxide monohydrate, na.ho.h2o, naoh water, water naoh, sodium hydroxide h2o, sodium hydroxide water, water sodium hydroxide, h2o naoh, naoh h2o, sodium,hydroxide,hydrate
PubChem CID 23719242
SMILES O.[OH-].[Na+]

Specifications

Color White
Boiling Point 1390°C
Melting Point 318.4°C
Assay Percent Range 99.996% (metals basis)
Density 2.13
Solubility Information Soluble in water and methanol.
Quantity 100 g
IUPAC Name sodium hydrate hydroxide
Formula Weight 58.01 (40.00 Anhydrous)
Odor Odorless
Physical Form Crystals
Chemical Name or Material Sodium hydroxide monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.