missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide (low chloride), ACS, 97.0% min

Product Code. 11346818
Change view
Click to view available options
Quantity:
2.5 g
100 g
500 g
2 kg
10 kg
2 x 12.5 kg
Unit Size:
100g
10kg
2.5g
25kg
2kg
500g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11346818 2 x 12.5 kg 25kg
11440320 2.5 g 2.5g
11366818 100 g 100g
11376818 500 g 500g
11356818 2 kg 2kg
11336818 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11346818 Supplier Thermo Scientific Alfa Aesar Supplier No. 013455.F6
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use in Chemical Manufacture, Rayon and Cellophane, As a Neutralizing Agent in Petroleum Refining | CAS: 1310-73-2 | HNaO | 39.997 g/mol

In chemical manufacture, rayon and cellophane, as a neutralizing agent in petroleum refining, in pulp, paper detergents and textiles, and as a lab reagent. NaOH solutions precipitate most metals as hydroxides from aqueous solutions of the metal saltsSodium hydroxide is widely used in variety of large scale applications such as pulp and paper manufacturing, in the production of soap and detergents as well as water treatment. It finds application in the base- driven hydrolysis of esters, amides and alkyl halides. It is also used as an industrial cleaning agent, an additive in drilling mud to increase alkalinity in bentonite in petroleum industry and as a powerful degreaser on stainless steel and glass bakeware. It is utilized in the Maxam-Gilbert DNA sequencing technique, chocolate and cocoa processing, soft drink processing, caramel coloring production and for thickening ice cream. It is often used to neutralize acids and prepare sodium salts of reagents. It forms a solution with sodium dodecyl sulfate and plays an important role for the decontamination of mycobacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Boiling Point 1390°C
Melting Point 318°C
Color White
Physical Form Pellets
Quantity 2 x 12.5 kg
Assay Percent Range 97.0% min.
Solubility Information Soluble in water,alcohol and glycerolSoluble in water,glycerol,ethanol and methanol. Insoluble in acetone and ether.
Formula Weight 40
Grade ACS
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Appearance White pellets
Vapor Pressure 3.5 hPa (3mm Hg) at 800°C
Density 2.13
Chemical Name or Material Sodium hydroxide, (low chloride)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.