missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogen carbonate, ACS, 99.7-100.3%

Product Code. 11357018
Change view
Click to view available options
Quantity:
500 g
2 kg
10 kg
Unit Size:
10kg
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357018 2 kg 2kg
11337018 500 g 500g
11347018 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11357018 Supplier Thermo Scientific Alfa Aesar Supplier No. 014707.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use As a Source of CO2, in Making Sodium Salts, Baking Powder | CAS: 144-55-8 | CHNaO3 | 84.01 g/mol

Sodium hydrogen carbonate is used as a laboratory chemical, a leavening agent, a neutralizer, as an active component in fire extinguishers, as a source of carbon dioxide and as a pH balancer in pools, as a cattle feed supplement and in spas and garden ponds. It is utilized for paint and corrosion removal, in baking powder, as a mild abrasive and a cleaning compound. In the field of medicine, it is used as an antacid to treat acid indigestion and heartburn.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
SMILES [Na+].OC([O-])=O

Especificaciones

Color White
Physical Form Granules
Quantity 2 kg
Assay Percent Range 99.7-100.3%
Beilstein 4153970
Solubility Information Soluble in water and methanol. Insoluble in ethanol.
IUPAC Name sodium;hydrogen carbonate
Formula Weight 84.01
Grade ACS
Odor Odorless
Refractive Index 1.5
Density 2.159
Chemical Name or Material Sodium hydrogen carbonate, For ACS analysis
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.