missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 13285393 Shop All J.T.Baker Products
Change view
Click to view available options
:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
13285393 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13285393 Supplier JT Baker Supplier No. 0348.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Exceeds ACS Specifications

Specifications

Boiling Point 1390°C (1013 hPa)
CAS 1310-73-2
Melting Point 323°C
Assay Percent Range min. 97.0%
Molecular Formula HNaO
MDL Number MFCD00003548
UN Number 1823
Density 2.13 g/cm³ (20°C)
Synonym Caustic soda
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 1 kg
SMILES [OH-].[Na+]
IUPAC Name sodium hydroxide
Molecular Weight (g/mol) 40.00
Packaging Plastic bottle for solids
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.