Learn More
Sodium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.5N, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| UN Number | UN1824 |
| Formula Weight | 40 |
| Odor | Odorless |
| Quantity | 6 U |
| Physical Form | Liquid |
| Chemical Name or Material | Sodium hydroxide Acculute Standard Volumetric Solution, Final Concentration 0.5N |
Frequently Asked Questions (FAQs)
The following is the dilution protocol:
- Transfer entire contents of one unit bottle into a 1L volumetric flask.
- Rinse bottle residue, the threads of the bottle, and inner bottle cap each with DI water, and transfer to the 1L flask.
- Repeat rinse three times. Bring the 1L flask to volume with DI water and mix well.
The final solution in the 1L flask has a concentration of 0.5N Sodium hydroxide.
1 unit of Sodium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.5N when diluted to 1 L will yield a final concentration of 0.5N Sodium hydroxide solution.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.