missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.1N, Thermo Scientific Chemicals

Product Code. 11329329
Change view
Click to view available options
Quantity:
1 U
6 U
Unit Size:
1 unit
6 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329329 1 U 1 unit
11339329 6 U 6 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329329 Supplier Thermo Scientific Alfa Aesar Supplier No. 088623.LQ
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

UN Number UN1824
Formula Weight 40
Odor Odorless
Quantity 1 U
Physical Form Liquid
Chemical Name or Material Sodium hydroxide Acculute Standard Volumetric Solution, Final Concentration 0.1N
What is the protocol for diluting 1 unit of Sodium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.1N (088623.LQ, 088623-LQ)?

The following is the dilution protocol:
- Transfer entire contents of 1 unit bottle into a 1L volumetric flask.
- Rinse bottle residue, the threads of the bottle, and inner bottle cap each with DI water, and transfer to the 1L flask.
- Repeat rinse three times. Bring the 1L flask to volume with DI water and mix well.
The final solution in the 1L flask has a concentration of 0.1N Sodium hydroxide.

What is 1 unit of Sodium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.1N (Cat. No. 088623.LQ, 088623-LQ)?

1 unit of Sodium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.1N, when diluted to 1 L, will yield a final concentration of 0.1N Sodium hydroxide solution.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.