missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 50% Solution, Water for Injection Quality Water, Biotech Reagent, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
19 L
Packaging:
Poly Bottle
Unit Size:
19 L
Specifications
Specifications
| CAS | 1310-73-2 |
| Molecular Formula | HNaO |
| Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
| InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
| Quantity | 19 L |
| SMILES | [OH-].[Na+] |
| IUPAC Name | sodium;hydroxide |
| Molecular Weight (g/mol) | 39.997 |
| PubChem CID | 14798 |
| ChEBI | CHEBI:32145 |
| Show More |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction