missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 1.0N Volumetric Solution, Macron Fine Chemicals™
Click to view available options
Quantity:
4 L
Unit Size:
4 L
Specifications
Specifications
| Color | Colorless |
| CAS | 1310-73-2 |
| Molecular Formula | HNaO |
| Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
| InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
| Quantity | 4 L |
| SMILES | [OH-].[Na+] |
| IUPAC Name | sodium;hydroxide |
| Molecular Weight (g/mol) | 39.997 |
| PubChem CID | 14798 |
| Show More |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction