missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 98%, Honeywell™

Product Code. 15610820 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15610820 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15610820 Supplier Honeywell Research Chemicals Supplier No. 062081KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

for the manufacture of mirrors, pellets, Cl <0.007%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Specifications

pH Alkaline
Color Colorless
Boiling Point 1390°C (1013 hPha)
CAS 1310-73-2
Melting Point 319°C
Molecular Formula HNaO
Linear Formula NaOH
MDL Number MFCD00003548
UN Number UN1823
Density 2.13g/cu.cm at 20°C
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Solubility Information Soluble in water, 1260g/L at 20°C
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 1 kg
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40g/mol
Percent Purity 98.0 to 102.0%
Odor Odorless
Packaging Plastic bottle
Vapor Density >1 (vs air)
Vapor Pressure 18mmHg (20°C), 3mmHg (37°C)
Physical Form Solid
Chemical Name or Material Sodium Hydroxide, For the manufacture of mirrors, pellets, Cl <0.001%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.