missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide 0.5M, BAKER ANALYZED, J.T. Baker™

Product Code. 15538344 Shop All J.T.Baker Products
Click to view available options
Quantity:
1 L
Packaging:
HDPE
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 15538344

Brand: JT Baker 5634.1000

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High quality grade volumetric solution is ready to use for laboratory and specialized industrial applications.

  • Standardized potentiometrically against an NIST Standard Reference Material, Potassium Hydrogen Phthalate
  • Label provides an actual Lot Analysis as a guarantee of purity and consistency
  • Analytical methods used for these determinations are based on standard test procedures described in technical publications
  • Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability

Specifications

Boiling Point 100°C
CAS 1310-73-2
Melting Point 0°C
Molecular Formula HNaO
Concentration 0.4975–0.5025mol/L
Density 1g/cm3
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Solubility Information Fully miscible in water
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 1 L
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 39.997g/mol
Grade Standardized potentiometrically against an NIST Standard Reference Material, Potassium Hydrogen Phthalate
Normality 0.5N
Packaging HDPE
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.