missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide 0.5 M (0.5 N) Analytical Volumetric Solution (AVL), Reagecon
Shop All Reagecon Products

Description
- Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
- Accurate, stable, reproducible and fully factorized.
- Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
- Stability is proven (either opened or unopened) for the shelf life of this product.
- Certified by Reagecon’s ISO 17025 Accredited Test Method (INAB Ref: 264T)
- Certified to a specification of ± 0.1%.
- Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
- May be used for manual titrations, or automatic titrations, to determine the end points by detection of any of several properties.
- Acid/base titrations, also called neutralization titrations, are used to determine either the amount of acid/base in a sample or the substance that can be converted to an acid/base.
Specifications
Specifications
| Concentration | 0.5 M (0.5 N) |
| Quantity | 1 L |
| Packaging | HDPE Bottle GL45 |
| Physical Form | Liquid |
Safety and Handling
Product Identifier
- SODIUM HYDROXIDE 0.5N (0.5M)
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- H314-Causes severe skin burns and eye damage.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: sodium hydroxide
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?