missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide 0.5 M (0.5 N) Analytical Volumetric Solution (AVL) Bag-in-Box, Reagecon

Product Code. 11532972 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
10 L
Unit Size:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11532972 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11532972 Supplier Reagecon Supplier No. SB205010
  • ONLINE EXCLUSIVE PRICE
    £160.00
    £117.43 / 10 L
    Save £42.57
    26% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Fully factorized, high purity, stable, certified product, developed and tested for acid/base titrations.

  • Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
  • Accurate, stable, reproducible and fully factorized.
  • Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
  • Stability is proven (either opened or unopened) for the shelf life of this product.
  • Certified by Reagecon’s ISO 17025 Accredited Test Method (INAB Ref: 264T)
  • Certified to a specification of ± 0.1%.
  • Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
  • May be used for manual titrations, or automatic titrations, to determine the end points by detection of any of several properties.
  • Acid/base titrations, also called neutralization titrations, are used to determine either the amount of acid/base in a sample or the substance that can be converted to an acid/base.

Specifications

Concentration 0.5M (0.5N)
Quantity 10 L
Packaging Bag-in-Box Container
Physical Form Liquid
compliance-icons
Product Identifier
  • SODIUM HYDROXIDE 0.5N (0.5M)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.