missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, 0.1N Standardized Solution, Thermo Scientific Chemicals

Catalog Number 11323747
Click to view available options
Quantity:
1 L
4 L
20 L
1310-73-2
HNaO
39.997
MFCD00003548
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
1310-73-2
HNaO
39.997
MFCD00003548
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]

Sodium hydroxide, 0.1N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 L
Formula Weight 40
Concentration or Composition (by Analyte or Components) Sodium hydroxide: 0.4%; Water: 99.6%
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Density 1.1g/cm3 at 20°C
Solubility Miscible with glycerol, ethanol and methanol; Immiscible with acetone and ether
Physical Form Liquid
Chemical Name or Material Sodium hydroxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'