missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide (0.1M in water (0.1N)), TRC

Product Code. 30411539
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
This item is not returnable. View return policy

Product Code. 30411539

Brand: Toronto Research Chemicals TRCS62419050ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium hydroxide
CAS 1310-73-2
Molecular Formula NaOH
Quantity 50 mL
Synonym Sodium hydroxide, Sodium hydroxide (Na(OH)), Sodium hydroxide (8CI), Aetznatron, Ascarite, BO-200B, Caustic Solution 70TW, Caustic soda, Collo-Grillrein, Collo-Tapetta, DL α, E 33, E 33 (degreasing agent), FC 4370, GR, GR (alkali reagent), MO 67, NSC 135799, Promoter 3308B, Reasol, Soda','caustic, StamperPrep, Tosoh Pearl, White caustic, ma-D 331
InChI Formula InChI=1S/Na.H2O/h;1H2/q+1;/p-1
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 40
Formula Weight 39.9925
Concentration or Composition (by Analyte or Components) 0.1 M
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.