missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide (0.1M in water (0.1N)), TRC
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Specifications
Specifications
| Chemical Name or Material | Sodium hydroxide |
| CAS | 1310-73-2 |
| Molecular Formula | NaOH |
| Quantity | 100 mL |
| Synonym | Sodium hydroxide, Sodium hydroxide (Na(OH)), Sodium hydroxide (8CI), Aetznatron, Ascarite, BO-200B, Caustic Solution 70TW, Caustic soda, Collo-Grillrein, Collo-Tapetta, DL α, E 33, E 33 (degreasing agent), FC 4370, GR, GR (alkali reagent), MO 67, NSC 135799, Promoter 3308B, Reasol, Soda','caustic, StamperPrep, Tosoh Pearl, White caustic, ma-D 331 |
| InChI Formula | InChI=1S/Na.H2O/h;1H2/q+1;/p-1 |
| SMILES | [OH-].[Na+] |
| IUPAC Name | sodium;hydroxide |
| Molecular Weight (g/mol) | 40 |
| Formula Weight | 39.9925 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction