missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- For Laboratory, Research, or Manufacturing Use
- Protect from air to avoid absorption of carbon dioxide
- Standardization at 25°C traceable to NIST Standard Reference Material
- Filtered through a 0.2 micron filter
Specifications
Specifications
| CAS | 1310-73-2 |
| Molecular Formula | HNaO |
| MDL Number | MFCD00003548 |
| UN Number | 1824 |
| Density | 1 g/cm³ (25°C) |
| Synonym | Caustic soda |
| InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
| Quantity | 1 L |
| SMILES | [OH-].[Na+] |
| IUPAC Name | sodium hydroxide |
| Show More |
Safety and Handling
Product Identifier
- Sodium hydroxide 0.1 N
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: sodium hydroxide
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?