missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydrosulfite, Honeywell Fluka™

Product Code. 15681510 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
2 kg
Unit Size:
100g
1kg
2kg
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15681510 5 g 5g
15641510 100 g 100g
15671510 500 g 500g
15651510 1 kg 1kg
15661510 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15681510 Supplier Honeywell-Fluka Supplier No. 1579535G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7775-14-6
Molecular Formula Na2O4S2
Molecular Weight (g/mol) 174.096
MDL Number MFCD00011640
InChI Key JVBXVOWTABLYPX-UHFFFAOYSA-L
Synonym sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros
PubChem CID 24489
ChEBI CHEBI:66870
SMILES [O-]S(=O)S(=O)[O-].[Na+].[Na+]

Specifications

Quantity 5 g
Linear Formula NaO2SSO2Na
UN Number 1384
Solubility Information Soluble in water
Formula Weight 174.11g/mol
Grade Technical
Concentration or Composition (by Analyte or Components) 85%
Chemical Name or Material Sodium Hydrosulfite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.