missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydrosulfide Hydrate, TRC

Product Code. 30427872
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427872 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30427872 Supplier Toronto Research Chemicals Supplier No. TRCS6348205G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Hydrosulfide Hydrate
CAS 207683-19-0
Quantity 5 g
Molecular Formula HNaS(xH2O)
Synonym Sodium Sulfide (Na(SH)) Hydrate, Sodium Hydrogen Sulfide Hydrate
InChI Formula InChI=1S/Na.H2S/h;1H2/q+1;/p-1
SMILES [Na]S
IUPAC Name sulfanylsodium
Molecular Weight (g/mol) 56.06
Formula Weight 55.9697
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.