missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrosulfide hydrate, pure, flakes

Product Code. 10125320 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic Bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10125320 2.5 kg Plastic Bottle 2.5kg
10461301 25 g Glass bottle 25g
10782961 1 kg Glass bottle 1kg
10648314 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10125320 Supplier Thermo Scientific Acros Supplier No. 296200025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 207683-19-0 | HNaS | 56.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207683-19-0
Molecular Formula HNaS
Molecular Weight (g/mol) 56.06
MDL Number MFCD00149796
InChI Key HYHCSLBZRBJJCH-UHFFFAOYSA-M
Synonym sodium hydrosulfide hydrate, hnas.h2o, nahs.h2o, sodium sulfanide hydrate, sodium bisulfide monohydrate, sodium hydrosulphide hydrate, sodium hydro sulfide hydrate, sodium hydrosulfide monohydrate, sodium hydrate sulfanide, sodium hydrosulfide hydrate, cp
PubChem CID 16211346
IUPAC Name sodium sulfanide
SMILES [Na+].[SH-]

Specifications

Melting Point 55.0°C
Physical Form Deliquescing Flakes
Quantity 2.5 kg
Linear Formula NaSH·xH2O
Fieser 03,266
Flash Point 90°C
Merck Index 14,8587
Solubility Information Solubility in water: 620g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 56.06
Percent Purity ≥67% min. (NaHS) typically 70 to 72%
Grade Pure
Odor Obnoxious
Packaging Plastic Bottle
Chemical Name or Material Sodium hydrosulfide hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.