missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrosulfide hydrate, pure, flakes

Product Code. 10782961 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic Bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
25g
This item is not returnable. View return policy

Product Code. 10782961

Brand: Thermo Scientific Acros 296200010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 207683-19-0 | HNaS | 56.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207683-19-0
Molecular Formula HNaS
Molecular Weight (g/mol) 56.06
MDL Number MFCD00149796
InChI Key HYHCSLBZRBJJCH-UHFFFAOYSA-M
Synonym sodium hydrosulfide hydrate, hnas.h2o, nahs.h2o, sodium sulfanide hydrate, sodium bisulfide monohydrate, sodium hydrosulphide hydrate, sodium hydro sulfide hydrate, sodium hydrosulfide monohydrate, sodium hydrate sulfanide, sodium hydrosulfide hydrate, cp
PubChem CID 16211346
IUPAC Name sodium sulfanide
SMILES [Na+].[SH-]

Specifications

Melting Point 55.0°C
Physical Form Deliquescing Flakes
Quantity 1 kg
Linear Formula NaSH·xH2O
Fieser 03,266
Flash Point 90°C
Merck Index 14,8587
Solubility Information Solubility in water: 620g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 56.06
Percent Purity ≥67% min. (NaHS) typically 70 to 72%
Grade Pure
Odor Obnoxious
Packaging Glass bottle
Chemical Name or Material Sodium hydrosulfide hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.