missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogencyanamide, 98%, Thermo Scientific Chemicals

Product Code. 11955631 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11955631 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11955631 Supplier Thermo Scientific Acros Supplier No. 438050250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17292-62-5
Molecular Formula CHN2Na
Molecular Weight (g/mol) 64.02
MDL Number MFCD00066500
InChI Key MBEGFNBBAVRKLK-UHFFFAOYSA-N
Synonym sodium hydrogencyanamide, sodium acid cyanamide, sodium cyanamide, sodium cyanamidate, sodium cyanoazanide, cyanamide, monosodium salt, monosodium cyanamide, sodium hydrogen cyanamide, cyanamide, sodium salt 1:1, cyanamide monosodium salt
PubChem CID 4293993
SMILES [Na+].[NH-]C#N

Specifications

Melting Point >360.0°C
Color Tan to White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 98%
Solubility Information Solubility in water: 800g/L (20°C)
IUPAC Name sodium cyanoazanide
Formula Weight 64.02
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Sodium hydrogencyanamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium hydrogencyanamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.